Invitrogen
Manufacturer Code:PA577534
Catalog # PIPA577534
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)GDSPPGNGSGPRHIND corresponding to amino acid residues 366-381 of the human Nogo receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NgR; NgR1; Nogo receptor; nogo receptor; Nogo-66 receptor; Nogo-66 receptor; Nogo66 receptor; Nogo66 receptor-1; Reticulon-4 receptor; RTN4R Reticulon 4 Receptor Nogo-66 Receptor; UNQ330/PRO526
Gene Aliases: NGR; NgR1; NOGOR; RTN4R; UNQ330/PRO526
UniProt ID: (Human) Q9BZR6, (Rat) Q99M75, (Mouse) Q99PI8
Entrez Gene ID: (Human) 65078, (Rat) 113912, (Mouse) 65079
Molecular Function:
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.