Invitrogen
Manufacturer Code:PA547773
Catalog # PIPA547773
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to an 11 aa sequence unique to the N-terminus of human Nogo-C |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Foocen; Human NogoA; My043 protein; neurite growth inhibitor 220; Neurite outgrowth inhibitor; Neuroendocrine-specific protein; Neuroendocrine-specific protein C homolog; Nogo protein; NSP; reticulon 5; Reticulon-4; Reticulon-4 Foocen Neurite outgrowth inhibitor Nogo protein Neuroendocrine-specific protein NSP Neuroendocrine-specific protein C homolog RTN-x Reticulon-5 RTN4 KIAA0886 NOGO My043 SP1507; Reticulon-5; RTN-x
Gene Aliases: ASY; KIAA0886; My043; Nbla00271; Nbla10545; NI220/250; NOGO; NOGO-A; Nogo-B; Nogo-C; NOGOC; NSP; NSP-CL; RTN-X; RTN4; RTN4-A; RTN4-B1; RTN4-B2; RTN4-C; SP1507
UniProt ID: (Human) Q9NQC3
Entrez Gene ID: (Human) 57142
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.