Novus Biologicals
Manufacturer Code:NBP224950SS
Catalog # NBP224950SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 64-74 (RNRRHHRHPRG) of human Nogo-B receptor (NgBR) was used as immunogen GenBank no sp Q96E22.1 NGBR_HUMAN (Miao et al 2006). This immunogen sequence is within the 93 amino acid putative ectodomain of the NgBR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C6orf68MGC117249 chromosome 6 open reading frame 68 MGC:7199 MGC7199 NgBR nogo-B receptor Nuclear undecaprenyl pyrophosphate synthase 1 homolog nuclear undecaprenyl pyrophosphate synthase 1 homolog (S. cerevisiae); Cis-prenyltransferase subunit NgBR; dehydrodolichyl diphosphate syntase complex subunit NUS1; Dehydrodolichyl diphosphate synthase complex subunit NUS1; di-trans,poly-cis-decaprenylcistransferase; NgBR; Nogo-B receptor; Nuclear undecaprenyl pyrophosphate synthase 1 homolog; transport and golgi organization 14 homolog
Gene Aliases: C6orf68; MGC:7199; NGBR; NUS1; TANGO14
UniProt ID: (Human) B2RWQ4
Entrez Gene ID: (Human) 116150
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.