Novus Biologicals
Manufacturer Code:NBP169286
Catalog # NBP169286
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to Nogo The peptide sequence was selected from the middle region of Nogo. Peptide sequence FRIYKGVIQAIQKSDEGHPFRAYLESEVAISEELVQKYSNSALGHVNCTI. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Protein Aliases: ASY foocen Human NogoA KIAA0886 My043 protein Nbla00271 Nbla10545 neurite growth inhibitor 220 Neurite outgrowth inhibitor Neuroendocrine-specific protein Neuroendocrine-specific protein C homolog NI220/250 Nogo protein NOGO-A Nogo-B Nogo-C NOGOC NOGORTN4-B1 NSP NSP-CL reticulon 4 reticulon 5 reticulon-4 reticulon-5 RTN4-A RTN4-B2 RTN4-C RTN-x; Foocen; Human NogoA; My043 protein; neurite growth inhibitor 220; Neurite outgrowth inhibitor; Neuroendocrine-specific protein; Neuroendocrine-specific protein C homolog; Nogo protein; NSP; reticulon 5; Reticulon-4; Reticulon-5; RTN-x
Gene Aliases: ASY; KIAA0886; My043; Nbla00271; Nbla10545; NI220/250; NOGO; NOGO-A; Nogo-B; Nogo-C; NOGOC; NSP; NSP-CL; RTN-X; RTN4; RTN4-A; RTN4-B1; RTN4-B2; RTN4-C; SP1507
UniProt ID: (Human) Q9NQC3
Entrez Gene ID: (Human) 57142
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.