Invitrogen
Manufacturer Code:PA523084
Catalog # PIPA523084
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an C-terminal region of the human NODAL protein (within residues 200-347). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: early embryo mesoderm formation; HTX5 nodal homolog; Nodal; Nodal homolog; transgenic lethal 413.d
Gene Aliases: HTX5; NODAL; Tg.413d
UniProt ID: (Human) Q96S42, (Mouse) P43021
Entrez Gene ID: (Human) 4838, (Mouse) 18119
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.