Invitrogen
Manufacturer Code:PA521981
Catalog # PIPA521981
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 698 and 953 of NOD1 (Uniprot IDNo.Q9Y239) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CARD4 CLR7.1 NLRC1 NLR family CARD domain containing 1 caspase recruitment domain family member 4 caspase recruitment domain-containing protein 4 nucleotide-binding oligomerization domain leucine rich repeat and CARD domain containing 1 nucleotide-binding oligomerization domain-containing protein 1; caspase recruitment domain family, member 4; Caspase recruitment domain-containing protein 4; NLR family, CARD domain containing 1; nucleotide-binding oligomerization domain containing 1; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 1; Nucleotide-binding oligomerization domain-containing protein 1
Gene Aliases: CARD4; CLR7.1; NLRC1; NOD1
UniProt ID: (Human) Q9Y239
Entrez Gene ID: (Human) 10392
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.