Invitrogen
Manufacturer Code:PA517328
Catalog # PIPA517328
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues near the carboxy terminus of Nod1 protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: caspase recruitment domain 4; caspase recruitment domain family, member 4; Caspase recruitment domain-containing protein 4; NLR family CARD domain containing 1 caspase recruitment domain family member 4 caspase recruitment domain-containing protein 4 nucleotide-binding oligomerization domain leucine rich repeat and CARD domain containing 1 nucleotide-binding oligomerization domain-containing protein 1; NLR family, CARD domain containing 1; nucleotide-binding oligomerization domain containing 1; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 1; Nucleotide-binding oligomerization domain-containing protein 1
Gene Aliases: C230079P11; CARD4; CLR7.1; F830007N14Rik; NLRC1; NOD1; RGD1562269
UniProt ID: (Human) Q9Y239, (Mouse) Q8BHB0
Entrez Gene ID: (Human) 10392, (Mouse) 107607, (Rat) 500133
Molecular Function:
nucleic acid binding
transcription cofactor
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.