Invitrogen
Manufacturer Code:PA5103899
Catalog # PIPA5103899
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NKX61 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Drosophila NK transcription factor related gene family 6 locus 1 homeobox protein NK-6 homolog A Homeobox protein Nkx-6.1 homeodomain transcription factor Nkx6.1 NK homeo box family 6 member A NK homeobox family 6 A NK6 homeobox 1 NK6 transcription factor homolog A NK6 transcription factor related locus 1 NK6 transcription factor related locus 1 Nkx6.1 Nkx61 Nkx6-1 Nkx6a; Drosophila NK transcription factor related, gene family 6, locus 1; Homeobox protein NK-6 homolog A; Homeobox protein Nkx-6.1; homeodomain transcription factor Nkx6.1; NK homeo box, family 6, member A; NK homeobox, family 6, A; NK6 transcription factor homolog A; NK6 transcription factor related locus 1; NK6 transcription factor related, locus 1
Gene Aliases: NKX6-1; NKX6.1; Nkx61; NKX6A
UniProt ID: (Human) P78426, (Mouse) Q99MA9, (Rat) O35762
Entrez Gene ID: (Human) 4825, (Mouse) 18096, (Rat) 65193
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.