Invitrogen
Manufacturer Code:PA585768
Catalog # PIPA585768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of mouse Nkx2.5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cardiac-specific homeobox; cardiac-specific homeobox 1; Csx antibody NK2 homeobox 5 antibody Nkx2-5 antibody Nkx2.5 antibody tinman antibody; Drosophila NK2 transcription factor related, locus 5; Homeobox protein CSX; Homeobox protein NK-2 homolog E; homeobox protein NKX 2-5; Homeobox protein Nkx-2.5; NK2 transcription factor related locus 5; NK2 transcription factor related, locus 5; NKX 2-5; rNKx-2.5; tinman paralog
Gene Aliases: CHNG5; CSX; CSX1; HLHS2; Nkx-2.5; NKX2-5; NKX2.5; NKX2E; NKX4-1; tinman; VSD3
UniProt ID: (Human) P52952, (Rat) O35767, (Mouse) P42582
Entrez Gene ID: (Human) 1482, (Rat) 114109, (Mouse) 18091
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.