Invitrogen
Manufacturer Code:PA519138
Catalog # PIPA519138
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (PRAYSDPDPAKDPR) corresponding to the internal amino acids of NKX2-5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Cardiac-specific homeobox; cardiac-specific homeobox 1; Drosophila NK2 transcription factor related, locus 5; Homeobox protein CSX; Homeobox protein NK-2 homolog E; homeobox protein NKX 2-5; Homeobox protein Nkx-2.5; NK2 transcription factor related locus 5; NK2 transcription factor related, locus 5; NKX 2-5; NKX2-5 NK2 transcription factor related locus 5 (Drosophila) CHNG5 CSX CSX1 NKX2.5 NKX2E NKX4-1 NK2 transcription factor homolog E NK2 transcription factor related locus 5 cardiac-specific homeo box tinman paralog; rNKx-2.5; tinman paralog
Gene Aliases: CHNG5; CSX; CSX1; HLHS2; Nkx-2.5; NKX2-5; NKX2.5; NKX2E; NKX4-1; tinman; VSD3
UniProt ID: (Human) P52952, (Rat) O35767, (Mouse) P42582
Entrez Gene ID: (Human) 1482, (Rat) 114109, (Mouse) 18091
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.