Invitrogen
Manufacturer Code:PA598740
Catalog # PIPA598740
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Homeobox protein Nkx-2.3 protein (52-148AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Homeobox protein NK-2 homolog C; Homeobox protein Nkx-2.3; NK2 transcription factor related, locus 3
Gene Aliases: CSX3; NK2.3; NKX2-3; NKX2.3; NKX23; NKX2C; NKX4-3
UniProt ID: (Human) Q8TAU0
Entrez Gene ID: (Human) 159296
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.