Invitrogen
Manufacturer Code:PA540630
Catalog # PIPA540630
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human NKX2-3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Drosophila NK2 transcription factor related, locus 3; Homeobox protein NK-2 homolog 3; Homeobox protein NK-2 homolog C; Homeobox protein Nkx-2.3; NK2 transcription factor related, locus 3; Nkx2-C
Gene Aliases: CSX3; NK2.3; Nkx-2.3; NKX2-3; NKX2.3; NKX23; NKX2C; NKX4-3; tinman
UniProt ID: (Human) Q8TAU0, (Mouse) P97334
Entrez Gene ID: (Dog) 100684882, (Human) 159296, (Bovine) 539451, (Horse) 100070479, (Rabbit) 103351415, (Mouse) 18089, (Guinea pig) 100724809, (Rat) 309389
Molecular Function: DNA binding protein helix-turn-helix transcription factor homeobox transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.