Proteintech
Manufacturer Code:269831AP150UL
Catalog # 50-173-5525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Nir2 Fusion Protein Ag25357 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DRES9 NIR 2 NIR2 PITPNM PITPnm 1 PITPNM1 Rd9 RDGB RDGB1 RDGBA RDGBA1; Drosophila retinal degeneration B homolog; Membrane-associated phosphatidylinositol transfer protein 1; NIR-2; phosphatidylinositol transfer protein, membrane associated 1; Phosphatidylinositol transfer protein, membrane-associated 1; PITPnm 1; PYK2 N-terminal domain-interacting receptor 2; retinal degeneration B alpha 1
Gene Aliases: DRES9; NIR2; PITPNM; PITPNM1; Rd9; RDGB; RDGB1; RDGBA; RDGBA1
UniProt ID: (Human) Q9BZ73
Entrez Gene ID: (Human) 9600
Molecular Function:
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.