Abnova
Manufacturer Code:MAB1679
Catalog # 89-110-573
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | KT54 |
Immunogen | A synthetic peptide (conjugated with BSA) corresponding to mouse Nipbl. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C or -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rat monoclonal antibody raised against synthetic peptide of Nipbl.
For Research Use Only
Protein Aliases: 4921518A06Rik; 4933421G18Rik; C79399; Delangin homolog; Idn3; Nipped-B-like protein; SCC2 homolog
Gene Aliases: Idn3; Nipbl; Scc2
UniProt ID: (Mouse) Q6KCD5
Entrez Gene ID: (Mouse) 71175
Molecular Function: DNA binding protein chromatin/chromatin-binding protein nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.