Novus Biologicals
Manufacturer Code:NBP290210V
Catalog # NB038148
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 29 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human Niemann-Pick type C2 (NP_006423.1 Met1-Leu151). |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: EDDM1 epididymal secretory protein E1 He1 HE1epididymal protein 1 Human epididymis-specific protein 1 MGC1333 Niemann-Pick disease type C2 protein Niemann-Pick disease type C2 NP-C2 tissue-specific secretory protein; epididymal protein 1; Epididymal secretory protein E1; He1; Human epididymis-specific protein 1; Niemann-Pick disease type C2 protein; Niemann-Pick disease, type C2; NPC intracellular cholesterol transporter 2; tissue-specific secretory protein
Gene Aliases: EDDM1; HE1; NPC2
UniProt ID: (Human) P61916
Entrez Gene ID: (Human) 10577
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.