R&D Systems
Manufacturer Code:AF8644
Catalog # AF8644
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | <em>S. frugiperda</em> insect ovarian cell line <em>Sf</em>21-derived recombinant human Niemann-Pick Type C2/NPC2 Glu20-Leu151 Accession # P61916 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 12 months from date of receipt -20 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after opening. 6 months -20 degreesC under sterile conditions after opening. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EDDM1 Epididymal Protein 1 epididymal secretory protein E1 HE1 HE1epididymal protein 1 Human epididymis-specific protein 1 MGC1333 Niemann-Pick disease type C2 protein Niemann-Pick disease type C2 NiemannPick Type C2 NPC2 NP-C2 tissue-specific secretory protein; epididymal protein 1; Epididymal secretory protein E1; He1; Human epididymis-specific protein 1; Niemann-Pick disease type C2 protein; Niemann-Pick disease, type C2; NPC intracellular cholesterol transporter 2; tissue-specific secretory protein
Gene Aliases: EDDM1; HE1; NPC2
UniProt ID: (Human) P61916
Entrez Gene ID: (Human) 10577
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.