Novus Biologicals
Manufacturer Code:NB400148F
Catalog # NB400148F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to the C-terminal region of human Niemann-Pick C. [UniProt# O15118] |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ98532 Niemann-Pick C1 protein Niemann-Pick disease type C1 NPC NPC1; Niemann-Pick C1 protein; Niemann-Pick disease, type C1; NPC intracellular cholesterol transporter 1; truncated Niemann-Pick C1
Gene Aliases: NPC; NPC1
UniProt ID: (Human) O15118
Entrez Gene ID: (Human) 4864
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.