Novus Biologicals
Manufacturer Code:NBP120901
Catalog # NBP120901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-DYRLTWNSSRYEGVN corresponding to internal region according to NP_000741.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor, nicotinic, beta 4 (neuronal); cholinergic receptor nicotinic beta 4 cholinergic receptor nicotinic beta polypeptide 4 neuronal acetylcholine receptor subunit beta-4 neuronal nicotinic receptor beta 4 subunit; cholinergic receptor, nicotinic beta 4; cholinergic receptor, nicotinic, beta 4 (neuronal); cholinergic receptor, nicotinic, beta polypeptide 4; Neuronal acetylcholine receptor subunit beta-4; neuronal nicotinic receptor beta 4 subunit
Gene Aliases: CHRNB4
UniProt ID: (Human) Q8WXC8
Entrez Gene ID: (Human) 1143
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.