Novus Biologicals
Manufacturer Code:NBP2822890.1ML
Catalog # NB030218
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human Nicotinic Acetylcholine Receptor beta 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor, nicotinic, beta 2 (neuronal); beta2 human neuronal nicotinic acetylcholine receptor; cholinergic receptor nicotinic beta 2 (neuronal) cholinergic receptor nicotinic beta polypeptide 2 (neuronal) EFNL3 nAChRB2 neuronal acetylcholine receptor subunit beta-2 neuronal nicotinic acetylcholine receptor beta 2; cholinergic receptor, nicotinic beta 2; cholinergic receptor, nicotinic, beta 2 (neuronal); cholinergic receptor, nicotinic, beta polypeptide 2 (neuronal); Neuronal acetylcholine receptor subunit beta-2; neuronal nicotinic acetylcholine receptor beta 2
Gene Aliases: CHRNB2; EFNL3; nAChRB2
UniProt ID: (Human) P17787
Entrez Gene ID: (Human) 1141
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.