Novus Biologicals
Manufacturer Code:NBP29434120UL
Catalog # NB023237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 26-240 of human CHRNA9 (NP_060051.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: acetylcholine receptor, neuronal nicotinic, alpha-9 subunit; cholinergic receptor nicotinic alpha 9 cholinergic receptor nicotinic alpha polypeptide 9 HSA243342 MGC142109 MGC142135 NACHR alpha 9 NACHR alpha-9 NACHRA9acetylcholine receptor neuronal nicotinic alpha-9 subunit neuronal acetylcholine receptor protein alpha-9 subunit neuronal acetylcholine receptor subunit alpha-9 nicotinic acetylcholine receptor subunit alpha 9 Nicotinic acetylcholine receptor subunit alpha-9; cholinergic receptor, nicotinic alpha 9; cholinergic receptor, nicotinic, alpha 9 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 9; NACHR alpha 9; NACHR alpha-9; neuronal acetylcholine receptor protein, alpha-9 subunit; Neuronal acetylcholine receptor subunit alpha-9; nicotinic acetylcholine receptor subunit alpha 9; Nicotinic acetylcholine receptor subunit alpha-9
Gene Aliases: CHRNA9; HSA243342; NACHRA9
UniProt ID: (Human) Q9UGM1
Entrez Gene ID: (Human) 55584
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.