Novus Biologicals
Manufacturer Code:NBP299069AF647
Catalog # NB057729
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Nicotinic Acetylcholine R alpha 7/CHRNA7. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: a7 nicotinic acetylcholine receptor; a7 nicotinic acetylcholine receptor alpha 7 neuronal nicotinic acetylcholine receptor alpha-7 nicotinic cholinergic receptor subunit cholinergic receptor nicotinic alpha 7 cholinergic receptor nicotinic alpha polypeptide 7 CHRNA7-2 NACHRA7 neuronal acetylcholine receptor protein alpha-7 chain neuronal acetylcholine receptor subunit alpha-7; alpha 7 neuronal nicotinic acetylcholine receptor; alpha-7 nicotinic cholinergic receptor subunit; cholinergic receptor, nicotinic alpha 7; cholinergic receptor, nicotinic, alpha 7 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 7; neuronal acetylcholine receptor protein, alpha-7 chain; Neuronal acetylcholine receptor subunit alpha-7
Gene Aliases: CHRNA7; CHRNA7-2; NACHRA7
UniProt ID: (Human) P36544
Entrez Gene ID: (Human) 1139
Molecular Function:
GABA receptor
acetylcholine receptor
ion channel
ligand-gated ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.