Novus Biologicals
Manufacturer Code:NBP287917
Catalog # NB035539
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human Nicotinic Acetylcholine R alpha 3/CHRNA3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cholinergic receptor nicotinic alpha 3 cholinergic receptor nicotinic alpha polypeptide 3 LNCR2 MGC104879 NACHRA3 neuronal acetylcholine receptor subunit alpha-3 neuronal nicotinic acetylcholine receptor alpha3 subunit PAOD2; cholinergic receptor, nicotinic alpha 3; cholinergic receptor, nicotinic, alpha 3 (neuronal); cholinergic receptor, nicotinic, alpha polypeptide 3; Neuronal acetylcholine receptor subunit alpha-3; neuronal nicotinic acetylcholine receptor, alpha3 subunit
Gene Aliases: CHRNA3; LNCR2; NACHRA3; PAOD2
UniProt ID: (Human) P32297
Entrez Gene ID: (Human) 1136
Molecular Function: GABA receptor acetylcholine receptor ion channel ligand-gated ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.