Invitrogen
Manufacturer Code:PA5116471
Catalog # PIPA5116471
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to N-terminal Rat NHE-1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: APNH; APNH APNH1 AW554487 LIKNS Mir5122 mir-5122 Na(+)/H(+) antiporter amiloride-sensitive Na(+)/H(+) exchanger 1 Na+/H+ antiporter Na+/H+ amiloride sensitive Na-Li countertransporter NHE1 NHE-1 PPP1R143 protein phosphatase 1 regulatory subunit 143 Slc9a1 slow-wave epilepsy sodium/hydrogen exchanger 1 Solute carrier family 9 (sodium/hydrogen exchanger 1) antiporter Na+/H+ (amiloride sensitive) solute carrier family 9 (sodium/hydrogen exchanger) isoform 1 (antiporter Na+/H+ amiloride sensitive) solute carrier family 9 (sodium/hydrogen exchanger) member 1 solute carrier family 9 (sodium/hydrogen exchanger) member 1 (antiporter Na+/H+ amiloride sensitive) solute carrier family 9 member 1 solute carrier family 9 member A1 solute carrier family 9 member 1 solute carrier family 9 subfamily A (NHE1 cation proton antiporter 1) member 1 swe; Na(+)/H(+) antiporter, amiloride-sensitive; Na(+)/H(+) exchanger 1; Na+/H+ antiporter; Na+/H+, amiloride sensitive; Na-Li countertransporter; NHE-1; protein phosphatase 1, regulatory subunit 143; slow-wave epilepsy; Sodium/hydrogen exchanger 1; Solute carrier family 9 (sodium/hydrogen exchanger 1), antiporter, Na+/H+, (amiloride sensitive); solute carrier family 9 (sodium/hydrogen exchanger), isoform 1 (antiporter, Na+/H+, amiloride sensitive); solute carrier family 9 (sodium/hydrogen exchanger), member 1; solute carrier family 9 (sodium/hydrogen exchanger), member 1 (antiporter, Na+/H+, amiloride sensitive); Solute carrier family 9 member 1; solute carrier family 9, member 1; solute carrier family 9, subfamily A (NHE1, cation proton antiporter 1), member 1
Gene Aliases: APNH; APNH1; AW554487; LIKNS; mir-5122; Mir5122; NHE-1; NHE1; PPP1R143; SLC9A1; swe
UniProt ID: (Human) P19634, (Rat) P26431, (Mouse) Q61165
Entrez Gene ID: (Human) 6548, (Rat) 24782, (Mouse) 20544
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.