Abnova
Manufacturer Code:MAB5597
Catalog # 89-112-307
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MC192 |
Immunogen | Rat Ngfr. |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Mouse (P75 knock-out) monoclonal antibody raised against Ngfr.
For Research Use Only
Protein Aliases: CD271; Gp80-LNGFR; LNGFR; low affinity nerve growth factor receptor; Low affinity neurotrophin receptor p75NTR; Low-affinity nerve growth factor receptor; nerve growth factor receptor (TNFR superfamily, member 16); nerve growth factor receptor, fast; NGF receptor; p75; p75 ICD; p75NTR; RNNGFRR; Tumor necrosis factor receptor superfamily member 16
Gene Aliases: LNGFR; Ngfr; p75; p75NTR; RNNGFRR; Tnfrsf16
UniProt ID: (Rat) P07174
Entrez Gene ID: (Rat) 24596
Molecular Function: cytokine receptor receptor tumor necrosis factor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.