Invitrogen
Manufacturer Code:PA579198
Catalog # PIPA579198
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived mouse Elastase/ELANE/ELA2 recombit protein (Position: S27-N265). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: elastase 2, neutrophil; Elastase-2; Leukocyte elastase; Neutrophil elastase; Neutrophil elastase Elastase-2 Leukocyte elastase Elane Ela2
Gene Aliases: Ela2; Elane; F430011M15Rik; NE
UniProt ID: (Mouse) Q3UP87
Entrez Gene ID: (Rat) 299606, (Mouse) 50701
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.