R&D Systems
Manufacturer Code:MAB91673SP
Catalog # MAB91673SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 950312 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Elastase/ELA-2 Met1-Asn252 Accession # P08246 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
Protein Aliases: Bone marrow serine protease; Bone marrow serine protease EC 3.4.21 EC 3.4.21.37 ELA2 ELA2granulocyte-derived elastase ELANE elastase 2 neutrophil elastase neutrophil expressed Elastase-2 GE HLEelastase-2 HNE Human leukocyte elastase Leukocyte Elastase medullasin NE neutrophil elastase PMN elastase PMN-E polymorphonuclear elastase SCN1; elastase 2, neutrophil; Elastase-2; granulocyte-derived elastase; HLE; Human leukocyte elastase; leukocyte elastase; Medullasin; Neutrophil elastase; PMN elastase; polymorphonuclear elastase
Gene Aliases: ELA2; ELANE; GE; HLE; HNE; NE; PMN-E; SCN1
UniProt ID: (Human) P08246
Entrez Gene ID: (Human) 1991
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.