R&D Systems
Manufacturer Code:MAB4517SP
Catalog # MAB4517SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 887105 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Neutrophil Elastase/ELA2 Met1-Arg260 Accession # Q3UP87 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Bone marrow serine protease; Bone marrow serine protease EC 3.4.21 EC 3.4.21.37 ELA2 ELA2granulocyte-derived elastase ELANE elastase 2 neutrophil elastase neutrophil expressed Elastase-2 GE HLEelastase-2 HNE Human leukocyte elastase Leukocyte Elastase medullasin NE neutrophil elastase PMN elastase PMN-E polymorphonuclear elastase SCN1; elastase 2, neutrophil; Elastase-2; granulocyte-derived elastase; HLE; Human leukocyte elastase; leukocyte elastase; Medullasin; Neutrophil elastase; PMN elastase; polymorphonuclear elastase
Gene Aliases: ELA2; ELANE; GE; HLE; HNE; NE; PMN-E; SCN1
UniProt ID: (Human) P08246
Entrez Gene ID: (Human) 1991
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.