Novus Biologicals
Manufacturer Code:NBP261657
Catalog # NBP2-61657
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 6B6G6 |
Immunogen | Purified recombinant fragment of human ELANE (AA: 140-267) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: Bone marrow serine protease; Bone marrow serine protease EC 3.4.21 EC 3.4.21.37 ELA2granulocyte-derived elastase elastase 2 neutrophil elastase neutrophil expressed Elastase-2 GE HLEelastase-2 HNE Human leukocyte elastase leukocyte elastase medullasin NE neutrophil elastase PMN elastase PMN-E polymorphonuclear elastase SCN1; elastase 2, neutrophil; Elastase-2; granulocyte-derived elastase; HLE; Human leukocyte elastase; leukocyte elastase; Medullasin; Neutrophil elastase; PMN elastase; polymorphonuclear elastase
Gene Aliases: ELA2; ELANE; GE; HLE; HNE; NE; PMN-E; SCN1
UniProt ID: (Human) P08246
Entrez Gene ID: (Human) 1991
Molecular Function: hydrolase protease serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.