Invitrogen
Manufacturer Code:PA520363
Catalog # PIPA520363
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 23 amino acid peptide from near the amino terminus of human neurotrypsin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Brain-specific serine protease 3; BSSP-3; BSSP-3 BSSP3 MRT1 brain-specific serine protease 3 leydin neurotrypsin; Leydin; Motopsin; Neurotrypsin; peptidase, serine, 12 (neurotrypsin, motopsin); protease, serine, 12 (neurotrypsin, motopsin); protease, serine, 12 neurotrypsin (motopsin); Serine protease 12
Gene Aliases: BSSP-3; BSSP3; motopsin; MRT1; Nt; PRSS12
UniProt ID: (Human) P56730, (Rat) G3V801, (Mouse) O08762
Entrez Gene ID: (Human) 8492, (Rat) 85266, (Mouse) 19142
Molecular Function:
hydrolase
oxidase
oxidoreductase
protease
receptor
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.