Novus Biologicals
Manufacturer Code:NB10056472SS
Catalog # NB10056472T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acids 150-200 of human Neurotensin receptor 2 was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Levocabastine-sensitive neurotensin receptor; neurotensin receptor 2 neurotensin receptor type 2 neurotensin receptor type 2 NT-R-2 NTR2Levocabastine-sensitive neurotensin receptor; Neurotensin receptor type 2; neurotensin receptor, type 2; NT-R-2
Gene Aliases: NTR2; NTSR2
UniProt ID: (Human) O95665
Entrez Gene ID: (Human) 23620
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.