Invitrogen
Manufacturer Code:MA525877
Catalog # PIMA525877
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3C4 |
Immunogen | Full length human recombit protein of NPTN produced in HEK293T cell |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: glycoprotein 55; Glycoprotein 55/65; glycoprotein 65; GP55 GP65 SDFR1 SDR1 np55 np65 SDR-1 stromal cell derived factor receptor 1 stromal cell-derived receptor 1; gp55/65; Neuroplastin; SDR-1; stromal cell derived factor receptor 1; Stromal cell-derived receptor 1
Gene Aliases: GP55; GP65; np55; np65; NPTN; SDFR1; SDR1
UniProt ID: (Human) Q9Y639, (Rat) P97546
Entrez Gene ID: (Human) 27020, (Dog) 487637, (Rhesus monkey) 699627, (Rat) 56064
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.