Invitrogen
Manufacturer Code:PA584072
Catalog # PIPA584072
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human NPTN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GP55 GP65 np55 np65 SDFR1 SDR1; Neuroplastin; SDR-1; stromal cell derived factor receptor 1; Stromal cell-derived receptor 1
Gene Aliases: GP55; GP65; np55; np65; NPTN; SDFR1; SDR1
UniProt ID: (Human) Q9Y639
Entrez Gene ID: (Human) 27020
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.