R&D Systems
Manufacturer Code:AF7818
Catalog # AF7818
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Neuroplastin Gln29-Pro338 predicted Accession # P97300 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: AW554172 GP55 GP55Stromal cell-derived receptor 1 GP65SDR-1 GP66 MGC102805 neuroplastin np55 np65 NPTN SDFR1 SDFR1DKFZp686L2477 SDR1 SDR1stromal cell derived factor receptor 1; Neuroplastin; SDR-1; stromal cell derived factor receptor 1; Stromal cell-derived receptor 1
Gene Aliases: GP55; GP65; np55; np65; NPTN; SDFR1; SDR1
UniProt ID: (Human) Q9Y639
Entrez Gene ID: (Human) 27020
Molecular Function:
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.