Invitrogen
Manufacturer Code:PA577526
Catalog # PIPA577526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)REDQGGEWKHGR corresponding to amino acid residues 747 - 758 of human Neuropilin-2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neuropilin-2; Neuropilin-2 (extracellular); neuropilin-2a(17); neuropilin-2a(22); neuropilin-2b(0); receptor for VEGF165 and semaphorins class3; Vascular endothelial cell growth factor 165 receptor 2
Gene Aliases: 1110048P06Rik; Np-2; NP2; Npn-2; NPN2; NRP2; PRO2714; VEGF165R2
UniProt ID: (Human) Q7LBX6, (Mouse) O35375, (Rat) O35276
Entrez Gene ID: (Human) 8828, (Mouse) 18187, (Rat) 81527
Molecular Function:
apolipoprotein
cell adhesion molecule
enzyme modulator
extracellular matrix protein
hydrolase
membrane-bound signaling molecule
metalloprotease
oxidase
oxidoreductase
protease
receptor
serine protease
signaling molecule
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.