R&D Systems
Manufacturer Code:MAB2215
Catalog # MAB2215
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 257107 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Neuropilin-2 Arg21-Tyr855 Accession # NP_003863 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC126574 neuropilin 2 neuropilin-2 Neuropilin2 neuropilin-2a(22) neuropilin-2b(0) NP2 NPN 2 NPN2 PRO2714 receptor for VEGF165 and semaphorins class3 Vascular endothelial cell growth factor 165 receptor 2 vascular endothelial growth factor-165 receptor 2 VEGF1265R2 VEGF165R2neuropilin-2a(17); Neuropilin-2; neuropilin-2a(17); neuropilin-2a(22); neuropilin-2b(0); receptor for VEGF165 and semaphorins class3; Vascular endothelial cell growth factor 165 receptor 2
Gene Aliases: NP2; NPN2; NRP2; PRO2714; VEGF165R2
UniProt ID: (Human) Q7LBX6
Entrez Gene ID: (Human) 8828
Molecular Function: apolipoprotein cell adhesion molecule enzyme modulator extracellular matrix protein hydrolase membrane-bound signaling molecule metalloprotease oxidase oxidoreductase protease receptor serine protease signaling molecule transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.