Invitrogen
Manufacturer Code:PA526079
Catalog # PIPA526079
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 722-750 amino acids from the C-terminal region of human NRP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BDCA4 CD304 NP1 NRP VEGF165R neuropilin-1 transmembrane receptor vascular endothelial cell growth factor 165 receptor; CD304; Neuropilin-1; transmembrane receptor; Vascular endothelial cell growth factor 165 receptor
Gene Aliases: BDCA4; CD304; NP1; NRP; NRP1; VEGF165R
UniProt ID: (Human) O14786
Entrez Gene ID: (Human) 8829
Molecular Function:
apolipoprotein
cell adhesion molecule
enzyme modulator
extracellular matrix protein
hydrolase
membrane-bound signaling molecule
metalloprotease
oxidase
oxidoreductase
protease
receptor
serine protease
signaling molecule
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.