R&D Systems
Manufacturer Code:AF3870SP
Catalog # AF3870SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Neuropilin-1 Phe22-Lys644 Accession # NP_001019799 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: BDCA4 BDCA-4 CD304 CD304 antigen DKFZp686A03134 DKFZp781F1414 neuropilin 1 neuropilin-1 Neuropilin1 NP1 NRP NRP1 transmembrane receptor Vascular endothelial cell growth factor 165 receptor VEGF165R; CD304; Neuropilin-1; transmembrane receptor; Vascular endothelial cell growth factor 165 receptor
Gene Aliases: BDCA4; CD304; NP1; NRP; NRP1; VEGF165R
UniProt ID: (Human) O14786
Entrez Gene ID: (Human) 8829
Molecular Function:
apolipoprotein
cell adhesion molecule
enzyme modulator
extracellular matrix protein
hydrolase
membrane-bound signaling molecule
metalloprotease
oxidase
oxidoreductase
protease
receptor
serine protease
signaling molecule
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.