R&D Systems
Manufacturer Code:MAB009100
Catalog # MAB009100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 62865 |
Immunogen | Synthetic peptide containing human Neurophysin II Accession &num P01186 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: antidiuretic hormone; Arg-vasopressin; arginine vasopressin ARVP AVP AVP-NPII AVRP neurohypophyseal NP2 vasopressin-neurophysin 2-copeptin VP; Arginine-vasopressin; AVP-NPII; Copeptin; neurohypophyseal; Neurophysin 2; Neurophysin-II; prepro-arginine-vasopressin-neurophysin II; prepro-AVP-NP II; Vasopressin-neurophysin 2-copeptin; vasopressin-neurophysin II-copeptin
Gene Aliases: ADH; ARVP; AVP; AVP-NPII; AVRP; VP
UniProt ID: (Human) P01185
Entrez Gene ID: (Human) 551
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.