Novus Biologicals
Manufacturer Code:NBP293197100UL
Catalog # NB028281
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 20-164 of human AVP (NP_000481.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antidiuretic hormone; Arg-vasopressin; arginine vasopressin ARVP AVP-NPII AVRP neurohypophyseal vasopressin-neurophysin 2-copeptin VP; Arginine-vasopressin; AVP-NPII; Copeptin; neurohypophyseal; Neurophysin 2; Neurophysin-II; prepro-arginine-vasopressin-neurophysin II; prepro-AVP-NP II; Vasopressin-neurophysin 2-copeptin; vasopressin-neurophysin II-copeptin
Gene Aliases: ADH; ARVP; AVP; AVP-NPII; AVRP; VP
UniProt ID: (Human) P01185
Entrez Gene ID: (Human) 551
Molecular Function: neuropeptide peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.