Novus Biologicals
Manufacturer Code:NBP105166UV
Catalog # NBP105166UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 8 |
Immunogen | Synthetic human/rat NPY coupled to carrier with glutaraldehyde |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 170 kDa melanoma membrane-bound gelatinase DKFZp686G13158 DPPIV EC 3.4.21.- FAPA Fibroblast activation protein alpha fibroblast activation protein alpha Integral membrane serine protease seprase; C-flanking peptide of NPY; CPON; Neuropeptide tyrosine; Neuropeptide Y; NPY; prepro-neuropeptide Y; Pro-neuropeptide Y
Gene Aliases: NPY; PYY4
UniProt ID: (Human) P01303
Entrez Gene ID: (Human) 4852
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.