Invitrogen
Manufacturer Code:PA3303
Catalog # APS303
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues W(1) Y K P A A G P H H Y S V G R A S G L L S S F H R F P S T(29) of mouse NPB protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPPL7; L7C; mPPL7; Neuropeptide B; Neuropeptide B-23; Neuropeptide B-29; NPB23; NPB29; PPL7 PPNPB; prepro-NPB; preproneuropeptide B; Preproprotein L7; rPPL7
Gene Aliases: L7; NPB; PPL7; PPNPB
UniProt ID: (Human) Q8NG41, (Rat) Q8K4P2, (Mouse) Q8K4P1
Entrez Gene ID: (Human) 256933, (Rat) 259222, (Mouse) 208990
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.