Novus Biologicals
Manufacturer Code:NBP294246100UL
Catalog # NB023048
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 360-426 of human NMUR1 (NP_006047.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (FM-3) FM3 FM-3 G protein-coupled receptor 66 GPC-R GPR66G-protein coupled receptor 66 G-protein coupled receptor FM-3 neuromedin U receptor 1 neuromedin-U receptor 1 NMU1R NMU-R1; G protein-coupled receptor 66; G-protein coupled receptor 66; G-protein coupled receptor FM-3; Neuromedin-U receptor 1; NMU-R1
Gene Aliases: (FM-3); FM-3; FM3; GPC-R; GPR66; NMU1R; NMUR1
UniProt ID: (Human) Q9HB89
Entrez Gene ID: (Human) 10316
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.