Invitrogen
Manufacturer Code:PA555269
Catalog # PIPA555269
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human Neuromedin U-8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neuromedin precursor-related peptide 33; Neuromedin precursor-related peptide 36; Neuromedin-U; Neuromedin-U-25; NmU-25; NURP33; NURP36; prepro-NMU
Gene Aliases: NMU
UniProt ID: (Human) P48645
Entrez Gene ID: (Human) 10874
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.