Invitrogen
Manufacturer Code:PA136127
Catalog # 01-672-285
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic porcine neuromedin U-8 conjugated to BSA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neuromedin precursor-related peptide 33; Neuromedin precursor-related peptide 36; Neuromedin-U; Neuromedin-U-23; NmU-23; NURP33; NURP36
Gene Aliases: Nmu; NmU-25
UniProt ID: (Rat) P12760
Entrez Gene ID: (Chicken) 422748, (Rat) 63887
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.