Novus Biologicals
Manufacturer Code:NBP242200AF647
Catalog # NB242200647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S110-29 |
Immunogen | Fusion protein amino acids 730-848 (intracellular C-terminus) of rat Neuroligin-3. Mouse: 99% identity (118/119 amino acids identical). Human: 98% identity (116/119 amino acids identical) ~60% identity with Neuroligin-1. ~40% identity with Neuroligin-2. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ASPGX1 AUTSX1 Gliotactin homolog KIAA1480HNL3 neuroligin 3 neuroligin-3 NL3; Gliotactin homolog; Neuroligin-3
Gene Aliases: HNL3; KIAA1480; NL3; NLGN3
UniProt ID: (Human) Q9NZ94
Entrez Gene ID: (Human) 54413
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.