Novus Biologicals
Manufacturer Code:NBP289824100UL
Catalog # NB021117
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 242 |
Immunogen | Recombinant Human Neuroligin 3/NLGN3 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: ASPGX1 AUTSX1 Gliotactin homolog KIAA1480HNL3 neuroligin 3 neuroligin-3 NL3; Gliotactin homolog; Neuroligin-3
Gene Aliases: HNL3; KIAA1480; NL3; NLGN3
UniProt ID: (Human) Q9NZ94
Entrez Gene ID: (Human) 54413
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.