Invitrogen
Manufacturer Code:PA577523
Catalog # PIPA577523
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)DLGPRAYDRFPGDS corresponding to amino acid residues 657-670 of rat Neuroligin 2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neuroligin-2; Nlgn2; ortholog of human and rat neuroligin 2 NLGN2
Gene Aliases: Kiaa1366; NL2; Nlgn2
UniProt ID: (Rat) Q62888, (Mouse) Q69ZK9
Entrez Gene ID: (Rat) 117096, (Mouse) 216856
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.