Invitrogen
Manufacturer Code:PA572822
Catalog # PIPA572822
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | NLGN2 antibody for a 19 amino acid peptide near the carboxy terminus of human NLGN2. The immunogen is within amino acids 630 - 680 of NLGN2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: neuroligin 2 KIAA1366 NLGN2; Neuroligin-2; ortholog of human and rat neuroligin 2 NLGN2
Gene Aliases: KIAA1366; NL2; NLGN2
UniProt ID: (Human) Q8NFZ4, (Rat) Q62888, (Mouse) Q69ZK9
Entrez Gene ID: (Human) 57555, (Rat) 117096, (Mouse) 216856
Molecular Function: esterase hydrolase lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.