Invitrogen
Manufacturer Code:PA518668
Catalog # PIPA518668
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (NPPDTDIRDPGKK) corresponding to the internal amino acids of NLGN2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: neuroligin 2 KIAA1366 NLGN2; Neuroligin-2
Gene Aliases: KIAA1366; NLGN2
UniProt ID: (Human) Q8NFZ4, (Rat) Q62888
Entrez Gene ID: (Human) 57555, (Rat) 117096
Molecular Function: esterase hydrolase lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.