Invitrogen
Manufacturer Code:PA5115657
Catalog # PIPA5115657
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human NLGN2 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Kiaa1366 neuroligin 2 neuroligin-2 NL2 Nlgn2 ortholog of human and rat neuroligin 2 NLGN2; Neuroligin-2; ortholog of human and rat neuroligin 2 NLGN2
Gene Aliases: KIAA1366; NL2; NLGN2
UniProt ID: (Human) Q8NFZ4, (Rat) Q62888, (Mouse) Q69ZK9
Entrez Gene ID: (Human) 57555, (Rat) 117096, (Mouse) 216856
Molecular Function: esterase hydrolase lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.